Assay Details
| Gene Symbol: AGT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342547.1 | NP_001328757.1 | 2 - 3 | 686 | 113 | |
| NM_120126.5 | NP_568064.1 | 2 - 3 | 631 | 113 | ||
| GenBank mRNA | AK227118.1 | - | 2 - 3 | 493 | 113 | |
| AY054264.1 | - | 2 - 3 | 485 | 113 | ||
| BT002306.1 | - | 2 - 3 | 461 | 113 | ||
More Information
Set Membership: |
|

AGT2
Genomic Map