Assay Details
| Gene Symbol: AT4G39900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120153.3 | NP_568073.1 | 5 - 6 | 801 | 87 | |
| GenBank mRNA | AY050943.1 | - | 5 - 6 | 693 | 87 | |
| AY057651.1 | - | 5 - 6 | 799 | 87 | ||
| AY079397.1 | - | 5 - 6 | 535 | 87 | ||
| AY084970.1 | - | 5 - 6 | 693 | 87 | ||
More Information
Set Membership: |
|

AT4G39900
Genomic Map