Assay Details
Gene Symbol: AT4G40050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342566.1 | NP_001328204.1 | 4 - 5 | 1258 | 74 | |
NM_120170.3 | NP_568079.1 | 4 - 5 | 1264 | 74 | ||
GenBank mRNA | AY040014.1 | - | 4 - 5 | 1115 | 74 | |
AY133799.1 | - | 4 - 5 | 1004 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
