Assay Details
| Gene Symbol: AT4G40050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342566.1 | NP_001328204.1 | 5 - 6 | 1451 | 100 | |
| NM_120170.3 | NP_568079.1 | 5 - 6 | 1457 | 100 | ||
| GenBank mRNA | AY040014.1 | - | 5 - 6 | 1308 | 100 | |
| AY133799.1 | - | 5 - 6 | 1197 | 100 | ||
More Information
Set Membership: |
|

AT4G40050
Genomic Map