Assay Details
Gene Symbol: AT4G40050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120170.3 | NP_568079.1 | 6 - 7 | 1551 | 94 | |
GenBank mRNA | AY040014.1 | - | 6 - 7 | 1402 | 94 | |
AY133799.1 | - | 6 - 7 | 1291 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
