Assay Details
| Gene Symbol: HB16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120171.3 | NP_195716.1 | 2 - 3 | 743 | 101 | |
| GenBank mRNA | AF370620.1 | - | 2 - 3 | 433 | 101 | |
More Information
Set Membership: |
|

HB16
Genomic Map