Assay Details
| Gene Symbol: PTR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342576.1 | NP_001331271.1 | 2 - 3 | 341 | 147 | |
| NM_001342577.1 | NP_001331272.1 | 2 - 3 | 326 | 147 | ||
| NM_120196.4 | NP_195738.1 | 2 - 3 | 302 | 147 | ||
More Information
Set Membership: |
|

PTR5
Genomic Map