Assay Details
| Gene Symbol: SQD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120200.4 | NP_568085.2 | 11 - 12 | 1491 | 95 | |
| GenBank mRNA | AF454354.1 | - | 11 - 12 | 1259 | 95 | |
| BT005796.1 | - | 11 - 12 | 1259 | 95 | ||
| KJ138712.1 | - | 11 - 12 | 1259 | 95 | ||
More Information
Set Membership: |
|

SQD2
Genomic Map