Assay Details
| Gene Symbol: SQD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342579.1 | NP_001331107.1 | 9 - 10 | 1157 | 69 | |
| NM_120200.4 | NP_568085.2 | 9 - 10 | 1211 | 69 | ||
| GenBank mRNA | AF454354.1 | - | 9 - 10 | 979 | 69 | |
| BT005796.1 | - | 9 - 10 | 979 | 69 | ||
| KJ138712.1 | - | 9 - 10 | 979 | 69 | ||
More Information
Set Membership: |
|

SQD2
Genomic Map