Assay Details
| Gene Symbol: LAX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120202.5 | NP_195744.1 | 5 - 6 | 1247 | 59 | |
| NM_202990.2 | NP_974719.1 | 3 - 4 | 571 | 59 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LAX1| Gene Symbol: LAX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120202.5 | NP_195744.1 | 5 - 6 | 1247 | 59 | |
| NM_202990.2 | NP_974719.1 | 3 - 4 | 571 | 59 | ||
Set Membership: |
|