Assay Details
| Gene Symbol: LECRKA4.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120232.2 | NP_195774.1 | 1 | 2344 | 175 | |
| GenBank mRNA | AF361597.1 | - | 1 | 2243 | 175 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LECRKA4.1| Gene Symbol: LECRKA4.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120232.2 | NP_195774.1 | 1 | 2344 | 175 | |
| GenBank mRNA | AF361597.1 | - | 1 | 2243 | 175 | |
Set Membership: |
|