Assay Details
| Gene Symbol: BRCA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120241.4 | NP_195783.3 | 11 - 12 | 2443 | 131 | |
| GenBank mRNA | AJ488305.1 | - | 11 - 12 | 2248 | 131 | |
| Gene Symbol: BRCA2(IV) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160720.1 | NP_001154192.1 | 12 - 13 | 2221 | 131 | |
| NM_001340223.1 | NP_001328738.1 | 11 - 12 | 2376 | 131 | ||
| NM_001340224.1 | NP_001328737.1 | 11 - 12 | 2376 | 131 | ||
| NM_001340225.1 | NP_001328739.1 | 11 - 12 | 2376 | 131 | ||
| NM_116219.5 | NP_191913.3 | 11 - 12 | 2376 | 131 | ||
| GenBank mRNA | AJ488304.1 | - | 11 - 12 | 2236 | 131 | |
More Information
Set Membership: |
|

BRCA2B
Genomic Map