Assay Details
| Gene Symbol: BRCA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120241.4 | NP_195783.3 | 13 - 14 | 2633 | 146 | |
| GenBank mRNA | AJ488305.1 | - | 13 - 14 | 2438 | 146 | |
| Gene Symbol: BRCA2(IV) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160720.1 | NP_001154192.1 | 14 - 15 | 2411 | 146 | |
| NM_001340223.1 | NP_001328738.1 | 13 - 14 | 2566 | 146 | ||
| NM_001340224.1 | NP_001328737.1 | 13 - 14 | 2566 | 146 | ||
| NM_001340225.1 | NP_001328739.1 | 13 - 14 | 2566 | 146 | ||
| NM_116219.5 | NP_191913.3 | 13 - 14 | 2566 | 146 | ||
| GenBank mRNA | AJ488304.1 | - | 13 - 14 | 2426 | 146 | |
More Information
Set Membership: |
|

BRCA2B
Genomic Map