Assay Details
| Gene Symbol: BRCA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120241.4 | NP_195783.3 | 16 - 17 | 3151 | 72 | |
| GenBank mRNA | AJ488305.1 | - | 16 - 17 | 2956 | 72 | |
More Information
Set Membership: |
|

BRCA2B
Genomic Map