Assay Details
| Gene Symbol: BRCA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120241.4 | NP_195783.3 | 17 - 18 | 3220 | 85 | |
| GenBank mRNA | AJ488305.1 | - | 17 - 18 | 3025 | 85 | |
More Information
Set Membership: |
|

BRCA2B
Genomic Map