Assay Details
| Gene Symbol: AT5G01740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120252.2 | NP_195794.1 | 1 | 582 | 118 | |
| GenBank mRNA | AY075604.1 | - | 1 | 530 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G01740| Gene Symbol: AT5G01740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120252.2 | NP_195794.1 | 1 | 582 | 118 | |
| GenBank mRNA | AY075604.1 | - | 1 | 530 | 118 | |
Set Membership: |
|