Assay Details
| Gene Symbol: SAUR21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120261.4 | NP_195803.1 | 1 | 2595 | 59 | |
| GenBank mRNA | AK229397.1 | - | 1 | 2249 | 59 | |
| BT015367.1 | - | 1 | 1977 | 59 | ||
| BT015725.1 | - | 1 | 1961 | 59 | ||
More Information
Set Membership: |
|

SAUR21
Genomic Map