Assay Details
| Gene Symbol: AT5G02060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120284.3 | NP_195826.1 | 1 - 2 | 574 | 112 | |
| GenBank mRNA | AK175360.1 | - | 1 - 2 | 291 | 112 | |
| BT014938.1 | - | 1 - 2 | 354 | 112 | ||
More Information
Set Membership: |
|

AT5G02060
Genomic Map