Assay Details
| Gene Symbol: NDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120291.4 | NP_195833.1 | 10 - 11 | 1148 | 89 | |
| GenBank mRNA | BT025628.1 | - | 10 - 11 | 1038 | 89 | |
More Information
Set Membership: |
|

NDP1
Genomic Map