Assay Details
| Gene Symbol: NDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120291.4 | NP_195833.1 | 11 - 12 | 1198 | 137 | |
| GenBank mRNA | BT025628.1 | - | 11 - 12 | 1088 | 137 | |
More Information
Set Membership: |
|

NDP1
Genomic Map