Assay Details
| Gene Symbol: NDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120291.4 | NP_195833.1 | 7 - 8 | 930 | 75 | |
| GenBank mRNA | BT025628.1 | - | 7 - 8 | 820 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDP1| Gene Symbol: NDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120291.4 | NP_195833.1 | 7 - 8 | 930 | 75 | |
| GenBank mRNA | BT025628.1 | - | 7 - 8 | 820 | 75 | |
Set Membership: |
|