Assay Details
| Gene Symbol: Fes1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342648.1 | NP_001332564.1 | 2 - 3 | 284 | 78 | |
| NM_120293.3 | NP_195835.1 | 1 - 2 | 140 | 78 | ||
| GenBank mRNA | AK118831.1 | - | 1 - 2 | 103 | 78 | |
| BT005554.1 | - | 1 - 2 | 92 | 78 | ||
More Information
Set Membership: |
|

Fes1C
Genomic Map