Assay Details
| Gene Symbol: Fes1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036745.2 | NP_001031822.1 | 5 - 6 | 598 | 117 | |
| NM_001342648.1 | NP_001332564.1 | 6 - 7 | 797 | 117 | ||
| NM_120293.3 | NP_195835.1 | 5 - 6 | 653 | 117 | ||
| GenBank mRNA | AK118831.1 | - | 5 - 6 | 616 | 117 | |
| BT005554.1 | - | 5 - 6 | 605 | 117 | ||
More Information
Set Membership: |
|

Fes1C
Genomic Map