Assay Details
| Gene Symbol: EMB2730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120303.5 | NP_195845.2 | 3 - 4 | 705 | 70 | |
| GenBank mRNA | AK176383.1 | - | 3 - 4 | 638 | 70 | |
| BT010457.1 | - | 3 - 4 | 580 | 70 | ||
More Information
Set Membership: |
|

EMB2730
Genomic Map