Assay Details
| Gene Symbol: EMB2730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120303.5 | NP_195845.2 | 6 - 7 | 1124 | 70 | |
| GenBank mRNA | AK176383.1 | - | 6 - 7 | 1057 | 70 | |
| BT010457.1 | - | 6 - 7 | 999 | 70 | ||
More Information
Set Membership: |
|

EMB2730
Genomic Map