Assay Details
| Gene Symbol: NAK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120307.4 | NP_195849.1 | 3 - 4 | 877 | 71 | |
| NM_180424.3 | NP_850755.1 | 4 - 5 | 832 | 71 | ||
| GenBank mRNA | AK228878.1 | - | 3 - 4 | 711 | 71 | |
| AY086765.1 | - | 3 - 4 | 683 | 71 | ||
More Information
Set Membership: |
|

NAK
Genomic Map