Assay Details
| Gene Symbol: NAK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120307.4 | NP_195849.1 | 4 - 5 | 1015 | 88 | |
| NM_180424.3 | NP_850755.1 | 5 - 6 | 970 | 88 | ||
| GenBank mRNA | AK228878.1 | - | 4 - 5 | 849 | 88 | |
| AY086765.1 | - | 4 - 5 | 821 | 88 | ||
More Information
Set Membership: |
|

NAK
Genomic Map