Assay Details
| Gene Symbol: NAK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120307.4 | NP_195849.1 | 5 - 6 | 1136 | 72 | |
| NM_180424.3 | NP_850755.1 | 6 - 7 | 1091 | 72 | ||
| GenBank mRNA | AK228878.1 | - | 5 - 6 | 970 | 72 | |
| AY086765.1 | - | 5 - 6 | 942 | 72 | ||
More Information
Set Membership: |
|

NAK
Genomic Map