Assay Details
| Gene Symbol: ALG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120319.4 | NP_195861.2 | 3 - 4 | 735 | 113 | |
| GenBank mRNA | AY099540.1 | - | 3 - 4 | 641 | 113 | |
| BT002117.1 | - | 3 - 4 | 519 | 113 | ||
| KJ138711.1 | - | 3 - 4 | 519 | 113 | ||
More Information
Set Membership: |
|

ALG10
Genomic Map