Assay Details
| Gene Symbol: ALG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120319.4 | NP_195861.2 | 7 - 8 | 1482 | 94 | |
| GenBank mRNA | AY099540.1 | - | 7 - 8 | 1388 | 94 | |
| BT002117.1 | - | 7 - 8 | 1266 | 94 | ||
| KJ138711.1 | - | 7 - 8 | 1266 | 94 | ||
More Information
Set Membership: |
|

ALG10
Genomic Map