Assay Details
| Gene Symbol: RHL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120360.4 | NP_195902.1 | 1 - 2 | 634 | 65 | |
| GenBank mRNA | AJ297842.1 | - | 1 - 2 | 630 | 65 | |
| BT028966.1 | - | 1 - 2 | 530 | 65 | ||
More Information
Set Membership: |
|

RHL2
Genomic Map