Assay Details
| Gene Symbol: LCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036746.2 | NP_001031823.1 | 8 - 9 | 1076 | 63 | |
| NM_120362.4 | NP_568108.1 | 8 - 9 | 1098 | 63 | ||
| GenBank mRNA | AK317012.1 | - | 8 - 9 | 1018 | 63 | |
More Information
Set Membership: |
|

LCL1
Genomic Map