Assay Details
| Gene Symbol: UPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120366.4 | NP_195908.1 | 16 - 17 | 4718 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UPL4| Gene Symbol: UPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120366.4 | NP_195908.1 | 16 - 17 | 4718 | 85 | |
Set Membership: |
|