Assay Details
| Gene Symbol: UPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342678.1 | NP_001331961.1 | 6 - 7 | 1479 | 70 | |
| NM_120366.4 | NP_195908.1 | 6 - 7 | 1479 | 70 | ||
More Information
Set Membership: |
|

UPL4
Genomic Map