Assay Details
| Gene Symbol: EIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120406.5 | NP_195948.1 | 6 - 7 | 1473 | 111 | |
| GenBank mRNA | AF141203.1 | - | 6 - 7 | 1428 | 111 | |
More Information
Set Membership: |
|

EIN2
Genomic Map