Assay Details
| Gene Symbol: AT5G03310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120409.2 | NP_195951.1 | 1 | 322 | 95 | |
| GenBank mRNA | BT012110.1 | - | 1 | 322 | 95 | |
| BT015704.1 | - | 1 | 262 | 95 | ||
| Gene Symbol: ADK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342696.1 | NP_001330384.1 | 11 | 1775 | 95 | |
| NM_001342697.1 | NP_001330385.1 | 10 | 1928 | 95 | ||
| NM_120408.4 | NP_195950.1 | 11 | 1557 | 95 | ||
More Information
Set Membership: |
|

AT5G03310
Genomic Map