Assay Details
| Gene Symbol: ATMDAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120444.5 | NP_568125.1 | 4 - 5 | 374 | 95 | |
| GenBank mRNA | AF428317.1 | - | 4 - 5 | 311 | 95 | |
| AK227165.1 | - | 4 - 5 | 321 | 95 | ||
| AY057628.1 | - | 4 - 5 | 314 | 95 | ||
| AY142000.1 | - | 4 - 5 | 262 | 95 | ||
More Information
Set Membership: |
|

ATMDAR2
Genomic Map