Assay Details
| Gene Symbol: KDTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342729.1 | NP_001331079.1 | 9 - 10 | 1266 | 92 | |
| NM_001342730.1 | NP_001331078.1 | 9 - 10 | 1288 | 92 | ||
| NM_120458.3 | NP_195997.2 | 10 - 11 | 1179 | 92 | ||
| GenBank mRNA | AY065115.1 | - | 10 - 11 | 1114 | 92 | |
| AY128758.1 | - | 10 - 11 | 1083 | 92 | ||
| KJ138708.1 | - | 10 - 11 | 1083 | 92 | ||
More Information
Set Membership: |
|

KDTA
Genomic Map