Assay Details
| Gene Symbol: TRFL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120459.4 | NP_195998.2 | 2 - 3 | 1108 | 81 | |
| GenBank mRNA | AB493735.1 | - | 2 - 3 | 1005 | 81 | |
More Information
Set Membership: |
|

TRFL10
Genomic Map