Assay Details
| Gene Symbol: TRFL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342731.1 | NP_001331807.1 | 3 - 4 | 1171 | 79 | |
| NM_120459.4 | NP_195998.2 | 3 - 4 | 1184 | 79 | ||
| GenBank mRNA | AB493735.1 | - | 3 - 4 | 1081 | 79 | |
More Information
Set Membership: |
|

TRFL10
Genomic Map