Assay Details
| Gene Symbol: TFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120465.3 | NP_196004.1 | 1 - 2 | 288 | 109 | |
| GenBank mRNA | BT024828.1 | - | 1 - 2 | 212 | 109 | |
| U77674.1 | - | 1 - 2 | 227 | 109 | ||
More Information
Set Membership: |
|

TFL1
Genomic Map