Assay Details
| Gene Symbol: MLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203290.1 | NP_001190219.1 | 2 - 3 | 415 | 100 | |
| NM_120467.4 | NP_196006.1 | 1 - 2 | 531 | 100 | ||
| GenBank mRNA | AF360126.1 | - | 1 - 2 | 415 | 100 | |
| AY051027.1 | - | 1 - 2 | 372 | 100 | ||
More Information
Set Membership: |
|

MLS
Genomic Map