Assay Details
| Gene Symbol: AT5G03880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120469.7 | NP_568128.1 | 7 - 8 | 729 | 93 | |
| GenBank mRNA | AY054607.1 | - | 7 - 8 | 729 | 93 | |
| AY081486.1 | - | 7 - 8 | 702 | 93 | ||
More Information
Set Membership: |
|

AT5G03880
Genomic Map