Assay Details
| Gene Symbol: APC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127387.3 | NP_565433.1 | 1 - 2 | 406 | 95 | |
| GenBank mRNA | AK229538.1 | - | 1 - 2 | 281 | 95 | |
| BT024912.1 | - | 1 - 2 | 173 | 95 | ||
More Information
Set Membership: |
|

APC10
Genomic Map