Assay Details
| Gene Symbol: HBI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202619.2 | NP_001189548.1 | 5 - 6 | 1241 | 82 | |
| NM_127388.4 | NP_565434.1 | 5 - 6 | 1176 | 82 | ||
| NM_179645.3 | NP_849976.1 | 5 - 6 | 1287 | 82 | ||
| GenBank mRNA | AF412099.1 | - | 5 - 6 | 1026 | 82 | |
| AY078030.1 | - | 5 - 6 | 942 | 82 | ||
More Information
Set Membership: |
|

HBI1
Genomic Map