Assay Details
| Gene Symbol: AT2G18370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127394.5 | NP_179428.1 | 1 - 2 | 492 | 150 | |
| GenBank mRNA | AK229548.1 | - | 1 - 2 | 399 | 150 | |
| AY084369.1 | - | 1 - 2 | 395 | 150 | ||
More Information
Set Membership: |
|

AT2G18370
Genomic Map