Assay Details
| Gene Symbol: TTN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127396.3 | NP_179430.1 | 2 - 3 | 352 | 152 | |
| GenBank mRNA | AF486852.1 | - | 2 - 3 | 182 | 152 | |
| AY035009.1 | - | 2 - 3 | 309 | 152 | ||
| AY063048.1 | - | 2 - 3 | 182 | 152 | ||
| AY088331.1 | - | 2 - 3 | 352 | 152 | ||
More Information
Set Membership: |
|

TTN5
Genomic Map