Assay Details
| Gene Symbol: emb2444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127407.4 | NP_179441.1 | 4 - 5 | 408 | 64 | |
| GenBank mRNA | AY150466.1 | - | 4 - 5 | 306 | 64 | |
More Information
Set Membership: |
|

emb2444
Genomic Map