Assay Details
| Gene Symbol: HB21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127411.2 | NP_179445.1 | 2 - 3 | 528 | 72 | |
| GenBank mRNA | AB493553.1 | - | 2 - 3 | 425 | 72 | |
| BT031362.1 | - | 2 - 3 | 425 | 72 | ||
More Information
Set Membership: |
|

HB21
Genomic Map