Assay Details
| Gene Symbol: CHR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335605.1 | NP_001318246.1 | 3 - 4 | 756 | 125 | |
| NM_001335606.1 | NP_001324143.1 | 3 - 4 | 807 | 125 | ||
| NM_001335607.1 | NP_001324142.1 | 3 - 4 | 807 | 125 | ||
| NM_001335608.1 | NP_001324146.1 | 3 - 4 | 807 | 125 | ||
| NM_001335609.1 | NP_001324145.1 | 3 - 4 | 829 | 125 | ||
| NM_001335610.1 | NP_001324144.1 | 2 - 3 | 984 | 125 | ||
| NM_127432.2 | NP_179466.1 | 2 - 3 | 984 | 125 | ||
More Information
Set Membership: |
|

CHR8
Genomic Map